siRNA Details
Gene Symbol: SHROOM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020859.3 | NP_065910.3 | 8 | 6052 | ||
XM_005263163.4 | XP_005263220.1 | 7 | 10506 | |||
XM_005263164.4 | XP_005263221.1 | 7 | 5064 | |||
XM_005263165.4 | XP_005263222.1 | 7 | 6490 | |||
XM_006714282.3 | XP_006714345.1 | 8 | 5207 | |||
XM_011532157.2 | XP_011530459.2 | 7 | 5894 | |||
XM_011532158.2 | XP_011530460.1 | 9 | 5853 | |||
XM_011532159.1 | XP_011530461.1 | 8 | 6042 | |||
XM_011532160.1 | XP_011530462.1 | 6 | 5054 | |||
XM_011532161.2 | XP_011530463.1 | 7 | 5232 | |||
XM_017008488.1 | XP_016863977.1 | 8 | 5210 | |||
XM_017008489.1 | XP_016863978.1 | 8 | 5111 | |||
GenBank mRNA | AB040914.1 | - | 4 | 3251 | ||
AB055660.1 | - | 8 | 6053 | |||
AK128677.1 | - | 4 | 4554 | |||
BC007291.1 | - | 4 | 1413 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
