siRNA Details
Gene Symbol: SHROOM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020859.3 | NP_065910.3 | 7 | 5341 | ||
XM_005263163.4 | XP_005263220.1 | 6 | 9795 | |||
XM_005263164.4 | XP_005263221.1 | 6 | 4353 | |||
XM_005263165.4 | XP_005263222.1 | 6 | 5779 | |||
XM_006714282.3 | XP_006714345.1 | 7 | 4496 | |||
XM_011532157.2 | XP_011530459.2 | 6 | 5183 | |||
XM_011532158.2 | XP_011530460.1 | 8 | 5142 | |||
XM_011532159.1 | XP_011530461.1 | 7 | 5331 | |||
XM_011532160.1 | XP_011530462.1 | 5 | 4343 | |||
XM_011532161.2 | XP_011530463.1 | 6 | 4521 | |||
XM_017008488.1 | XP_016863977.1 | 7 | 4499 | |||
XM_017008489.1 | XP_016863978.1 | 7 | 4400 | |||
GenBank mRNA | AB040914.1 | - | 3 | 2540 | ||
AB055660.1 | - | 7 | 5342 | |||
AK128677.1 | - | 3 | 3843 | |||
BC007291.1 | - | 3 | 702 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
