siRNA Details
Gene Symbol: PPM1M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122870.2 | NP_001116342.1 | 9 | 1197 | ||
NM_144641.3 | NP_653242.3 | 10 | 1563 | |||
XM_005264879.2 | XP_005264936.1 | 10 | 1656 | |||
XM_005264880.3 | XP_005264937.1 | 9 | 1376 | |||
XM_005264881.1 | XP_005264938.1 | 10 | 1452 | |||
GenBank mRNA | AK056894.1 | - | 9 | 1799 | ||
AK096681.1 | - | 8 | 1491 | |||
AL832484.1 | - | 8 | 1471 | |||
BC009644.1 | - | 9 | 1158 | 3905720 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
