siRNA Details
Gene Symbol: PPM1M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122870.2 | NP_001116342.1 | 5 | 567 | ||
NM_144641.3 | NP_653242.3 | 6 | 933 | |||
XM_005264879.2 | XP_005264936.1 | 6 | 1001 | |||
XM_005264880.3 | XP_005264937.1 | 5 | 746 | |||
XM_005264881.1 | XP_005264938.1 | 6 | 822 | |||
GenBank mRNA | AB474372.1 | - | 6 | 901 | ||
AK056894.1 | - | 6 | 822 | |||
AK096681.1 | - | 5 | 925 | |||
AK129647.1 | - | 1 | 15 | |||
AL832484.1 | - | 4 | 841 | |||
BC009644.1 | - | 5 | 528 | 3905720 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
