siRNA Details
Gene Symbol: SUN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130965.2 | NP_001124437.1 | 1 | 34 | ||
NM_001171944.1 | NP_001165415.1 | 2 | 93 | |||
NM_001171945.1 | NP_001165416.1 | 3 | 313 | |||
NM_001171946.1 | NP_001165417.1 | 2 | 81 | |||
GenBank mRNA | AB018353.1 | - | 2 | 51 | ||
AB648918.1 | - | 1 | 14 | |||
AK022469.1 | - | 2 | 61 | |||
AK302896.1 | - | 2 | 65 | |||
AK309120.1 | - | 3 | 285 | |||
BC013613.1 | - | 2 | 81 | |||
BC142707.1 | - | 1 | 44 | 40148817 | ||
BC152418.1 | - | 1 | 51 | 8862528 | ||
BX538211.1 | - | 2 | 62 | |||
DA112466.1 | - | 2 | 93 | |||
DA666453.1 | - | 2 | 93 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
