siRNA Details
Gene Symbol: C7orf34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178829.4 | NP_849151.2 | 2 | 443 | ||
GenBank mRNA | AB083484.1 | - | 2 | 391 | ||
AY927625.1 | - | 1 | 667 | |||
BC014596.1 | - | 2 | 448 | 4825428 | ||
KU178863.1 | - | 2 | 327 | |||
KU178864.1 | - | 1 | 914 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
