siRNA Details
Gene Symbol: WAPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318328.1 | NP_001305257.1 | 18 | 3880 | ||
NM_015045.3 | NP_055860.1 | 18 | 3898 | |||
XM_011539548.1 | XP_011537850.1 | 18 | 3948 | |||
XM_017015979.1 | XP_016871468.1 | 17 | 3795 | |||
XM_017015980.1 | XP_016871469.1 | 17 | 3738 | |||
XM_017015981.1 | XP_016871470.1 | 15 | 3485 | |||
GenBank mRNA | AB065003.1 | - | 18 | 3901 | ||
AF479418.1 | - | 19 | 3576 | |||
AK000849.1 | - | 7 | 868 | |||
AK027005.1 | - | 11 | 1621 | |||
BC017393.1 | - | 10 | 1169 | |||
BC140858.1 | - | 18 | 3604 | 9021726 | ||
BC150269.1 | - | 19 | 3749 | 9007232 | ||
D87450.1 | - | 19 | 3749 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
