siRNA Details
Gene Symbol: WAPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318328.1 | NP_001305257.1 | 19 | 4005 | ||
NM_015045.3 | NP_055860.1 | 19 | 4023 | |||
XM_011539548.1 | XP_011537850.1 | 19 | 4073 | |||
XM_017015979.1 | XP_016871468.1 | 18 | 3920 | |||
XM_017015980.1 | XP_016871469.1 | 18 | 3863 | |||
XM_017015981.1 | XP_016871470.1 | 16 | 3610 | |||
GenBank mRNA | AB065003.1 | - | 19 | 4026 | ||
AF479418.1 | - | 20 | 3701 | |||
AK000849.1 | - | 8 | 993 | |||
AK027005.1 | - | 12 | 1746 | |||
BC017393.1 | - | 11 | 1294 | |||
BC140858.1 | - | 19 | 3729 | 9021726 | ||
BC150269.1 | - | 20 | 3874 | 9007232 | ||
D87450.1 | - | 20 | 3874 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
