siRNA Details
Gene Symbol: ISCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272007.1 | NP_001258936.1 | 3 | 260 | ||
NM_194279.3 | NP_919255.2 | 4 | 376 | |||
GenBank mRNA | AK290728.1 | - | 4 | 349 | ||
BC032893.2 | - | 4 | 331 | 5259721 | ||
BX114918.1 | - | 4 | 433 | |||
BX248252.1 | - | 4 | 334 | |||
DB501282.1 | - | 3 | 215 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
