siRNA Details
Gene Symbol: ISCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194279.3 | NP_919255.2 | 3 | 325 | ||
GenBank mRNA | AK290728.1 | - | 3 | 298 | ||
BC015771.1 | - | 3 | 285 | 4849790 | ||
BC032893.2 | - | 3 | 280 | 5259721 | ||
BX114918.1 | - | 3 | 382 | |||
BX248252.1 | - | 3 | 283 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
