siRNA Details
Gene Symbol: TTC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317037.1 | NP_001303966.1 | 13 | 1863 | ||
NM_144965.2 | NP_659402.1 | 13 | 1902 | |||
XM_005251733.1 | XP_005251790.1 | 12 | 1714 | |||
XM_005251734.1 | XP_005251791.1 | 12 | 1745 | |||
XM_006716970.1 | XP_006717033.1 | 13 | 1862 | |||
XM_006716971.1 | XP_006717034.1 | 12 | 1689 | |||
XM_011518284.1 | XP_011516586.1 | 12 | 1758 | |||
XM_011518285.1 | XP_011516587.1 | 11 | 1615 | |||
XM_011518288.1 | XP_011516590.1 | 10 | 1473 | |||
XM_011518289.2 | XP_011516591.1 | 13 | 1786 | |||
XM_011518290.1 | XP_011516592.1 | 12 | 1733 | |||
XM_011518291.1 | XP_011516593.1 | 12 | 1787 | |||
XM_011518292.1 | XP_011516594.1 | 11 | 1658 | |||
XM_011518293.1 | XP_011516595.1 | 13 | 1900 | |||
GenBank mRNA | AK057342.1 | - | 13 | 1887 | ||
AK074105.1 | - | 6 | 3252 | |||
AK302737.1 | - | 13 | 1848 | |||
BC031281.1 | - | 13 | 1900 | 5295890 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
