siRNA Details
Gene Symbol: TTC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317037.1 | NP_001303966.1 | 11 | 1521 | ||
NM_144965.2 | NP_659402.1 | 11 | 1560 | |||
XM_005251733.1 | XP_005251790.1 | 10 | 1372 | |||
XM_005251734.1 | XP_005251791.1 | 10 | 1403 | |||
XM_006716970.1 | XP_006717033.1 | 11 | 1520 | |||
XM_006716971.1 | XP_006717034.1 | 10 | 1347 | |||
XM_011518283.1 | XP_011516585.1 | 11 | 1558 | |||
XM_011518284.1 | XP_011516586.1 | 10 | 1416 | |||
XM_011518289.2 | XP_011516591.1 | 11 | 1444 | |||
XM_011518290.1 | XP_011516592.1 | 10 | 1391 | |||
XM_011518293.1 | XP_011516595.1 | 11 | 1558 | |||
XM_011518294.2 | XP_011516596.1 | 11 | 1558 | |||
XM_011518295.1 | XP_011516597.1 | 11 | 1558 | |||
GenBank mRNA | AK057342.1 | - | 11 | 1545 | ||
AK074105.1 | - | 5 | 796 | |||
AK302737.1 | - | 11 | 1506 | |||
BC031281.1 | - | 11 | 1558 | 5295890 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
