siRNA Details
Gene Symbol: SLC27A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198580.2 | NP_940982.1 | 12 | 2262 | ||
XM_011528000.1 | XP_011526302.1 | 14 | 2439 | |||
XM_011528001.2 | XP_011526303.1 | 13 | 2238 | |||
XM_011528002.2 | XP_011526304.1 | 14 | 2219 | |||
XM_011528003.2 | XP_011526305.1 | 13 | 2432 | |||
XM_017026781.1 | XP_016882270.1 | 14 | 2376 | |||
GenBank mRNA | AK131101.1 | - | 6 | 3008 | ||
AK296355.1 | - | 12 | 2180 | |||
AK299852.1 | - | 13 | 2531 | |||
BC013149.1 | - | 6 | 693 | |||
BC059399.1 | - | 12 | 2215 | 30341529 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
