siRNA Details
Gene Symbol: SLC27A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198580.2 | NP_940982.1 | 2 | 290 | ||
XM_011528000.1 | XP_011526302.1 | 3 | 431 | |||
XM_011528001.2 | XP_011526303.1 | 2 | 230 | |||
XM_011528002.2 | XP_011526304.1 | 3 | 211 | |||
XM_011528003.2 | XP_011526305.1 | 3 | 460 | |||
XM_017026781.1 | XP_016882270.1 | 3 | 368 | |||
GenBank mRNA | AK294238.1 | - | 3 | 436 | ||
AK296355.1 | - | 2 | 208 | |||
AK299852.1 | - | 3 | 485 | |||
AK315935.1 | - | 2 | 196 | |||
BC059399.1 | - | 2 | 243 | 30341529 | ||
CN333611.1 | - | 2 | 292 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
