siRNA Details
Gene Symbol: PIEZO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142864.3 | NP_001136336.2 | 48 | 7207 | ||
GenBank mRNA | AB161230.1 | - | 37 | 5658 | ||
BC008073.2 | - | 7 | 796 | |||
BC150271.1 | - | 37 | 5495 | 9007243 | ||
D87071.1 | - | 37 | 5495 | |||
KC602455.1 | - | 48 | 6948 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
