siRNA Details
Gene Symbol: IRX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134222.1 | NP_001127694.1 | 5 | 1854 | ||
NM_033267.4 | NP_150366.1 | 4 | 2608 | |||
GenBank mRNA | AY335940.1 | - | 5 | 1622 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
