siRNA Details
Gene Symbol: SESN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199933.1 | NP_001186862.1 | 10 | 2357 | ||
NM_001199934.1 | NP_001186863.1 | 10 | 2399 | |||
NM_014454.2 | NP_055269.1 | 10 | 2871 | |||
GenBank mRNA | AF033120.1 | - | 10 | 2350 | ||
AF033121.1 | - | 10 | 2399 | |||
AF033122.1 | - | 10 | 2450 | |||
AJ420599.1 | - | 1 | 873 | |||
AK001886.1 | - | 5 | 1353 | |||
AL133559.1 | - | 1 | 665 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
