siRNA Details
Gene Symbol: SESN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199933.1 | NP_001186862.1 | 9 | 1393 | ||
NM_001199934.1 | NP_001186863.1 | 9 | 1435 | |||
NM_014454.2 | NP_055269.1 | 9 | 1907 | |||
GenBank mRNA | AF033120.1 | - | 9 | 1386 | ||
AF033121.1 | - | 9 | 1435 | |||
AF033122.1 | - | 9 | 1486 | |||
AK001886.1 | - | 4 | 389 | |||
AK290157.1 | - | 9 | 1714 | |||
AK290551.1 | - | 9 | 1390 | |||
BC112036.1 | - | 9 | 1539 | 8327504 | ||
BC113569.1 | - | 9 | 1527 | 8322621 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
