siRNA Details
Gene Symbol: LETM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012318.2 | NP_036450.1 | 3 | 632 | ||
XM_006713884.1 | XP_006713947.1 | 3 | 340 | |||
XM_017008187.1 | XP_016863676.1 | 2 | 753 | |||
GenBank mRNA | AF061025.1 | - | 3 | 632 | ||
AK293572.1 | - | 3 | 340 | |||
AL133650.1 | - | 2 | 398 | |||
BC014500.1 | - | 3 | 532 | 4860194 | ||
BC021208.1 | - | 3 | 557 | 4126510 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
