siRNA Details
Gene Symbol: WAC-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033805.1 | 3 | 2010 | |||
GenBank mRNA | AK001143.1 | - | 1 | 1215 | ||
BC045818.1 | - | 3 | 1985 |
Gene Symbol: RPL7L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198486.3 | NP_940888.3 | 7 | 1208 | ||
NR_134562.1 | 7 | 1324 | ||||
NR_134563.1 | 6 | 1102 | ||||
XM_017010780.1 | XP_016866269.1 | 6 | 2350 | |||
GenBank mRNA | BC058020.1 | - | 7 | 899 | 6736635 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
