siRNA Details
Gene Symbol: RALGAPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282917.1 | NP_001269846.1 | 13 | 2136 | ||
NM_001282918.1 | NP_001269847.1 | 13 | 2278 | |||
NM_020336.3 | NP_065069.1 | 13 | 2278 | |||
XM_005260462.2 | XP_005260519.1 | 14 | 2272 | |||
XM_005260463.2 | XP_005260520.1 | 14 | 2272 | |||
XM_005260464.2 | XP_005260521.1 | 14 | 2272 | |||
XM_005260465.2 | XP_005260522.1 | 13 | 2236 | |||
XM_017027966.1 | XP_016883455.1 | 14 | 2272 | |||
XM_017027967.1 | XP_016883456.1 | 13 | 2236 | |||
XM_017027968.1 | XP_016883457.1 | 13 | 1798 | |||
XM_017027969.1 | XP_016883458.1 | 14 | 2301 | |||
GenBank mRNA | AB033045.3 | - | 13 | 2268 | ||
AK123576.1 | - | 12 | 1937 | |||
AK125260.1 | - | 1 | 39 | |||
AK125916.1 | - | 8 | 1236 | |||
AL834436.1 | - | 11 | 1500 | |||
BC047482.1 | - | 13 | 2241 | 5270777 | ||
CR936800.1 | - | 13 | 2042 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
