siRNA Details
Gene Symbol: MTCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015210.3 | NP_056025.2 | 9 | 2189 | ||
XM_005258098.3 | XP_005258155.1 | 7 | 3089 | |||
XM_005258099.4 | XP_005258156.1 | 7 | 3089 | |||
XM_011525640.2 | XP_011523942.1 | 9 | 3290 | |||
XM_011525641.2 | XP_011523943.1 | 8 | 3212 | |||
XM_017025671.1 | XP_016881160.1 | 9 | 2659 | |||
XM_017025672.1 | XP_016881161.1 | 9 | 2260 | |||
XM_017025673.1 | XP_016881162.1 | 10 | 2267 | |||
XM_017025674.1 | XP_016881163.1 | 9 | 2314 | |||
XM_017025675.1 | XP_016881164.1 | 9 | 2189 | |||
GenBank mRNA | AB018345.2 | - | 7 | 2106 | ||
AK131528.1 | - | 9 | 2189 | |||
BC040542.1 | - | 2 | 86 | 5268201 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
