siRNA Details
Gene Symbol: MTCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015210.3 | NP_056025.2 | 17 | 5274 | ||
XM_005258098.3 | XP_005258155.1 | 14 | 6117 | |||
XM_005258099.4 | XP_005258156.1 | 15 | 6174 | |||
XM_011525640.2 | XP_011523942.1 | 17 | 6348 | |||
XM_011525641.2 | XP_011523943.1 | 16 | 6297 | |||
XM_017025671.1 | XP_016881160.1 | 17 | 5801 | |||
XM_017025672.1 | XP_016881161.1 | 17 | 5402 | |||
XM_017025673.1 | XP_016881162.1 | 18 | 5409 | |||
XM_017025674.1 | XP_016881163.1 | 17 | 5456 | |||
XM_017025675.1 | XP_016881164.1 | 17 | 5331 | |||
GenBank mRNA | AB018345.2 | - | 15 | 5191 | ||
AK131528.1 | - | 17 | 5274 | |||
BC040542.1 | - | 10 | 3144 | 5268201 | ||
BX648455.1 | - | 1 | 1933 | |||
BX648911.1 | - | 1 | 2037 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
