siRNA Details
Gene Symbol: TMEM37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183240.2 | NP_899063.2 | 2 | 111 | ||
XM_005263598.4 | XP_005263655.1 | 2 | 411 | |||
XM_006712300.3 | XP_006712363.1 | 2 | 235 | |||
XM_011510659.2 | XP_011508961.1 | 2 | 128 | |||
GenBank mRNA | AF361356.1 | - | 2 | 77 | ||
BC046362.1 | - | 1 | 236 | 5221396 | ||
DR003856.1 | - | 2 | 118 | |||
DR005637.1 | - | 2 | 100 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
