siRNA Details
Gene Symbol: DENND4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017925.6 | NP_060395.5 | 25 | 4696 | ||
NR_073201.3 | 25 | 4562 | ||||
XM_006716804.3 | XP_006716867.1 | 26 | 4811 | |||
XM_006716805.3 | XP_006716868.1 | 26 | 4811 | |||
XM_006716807.3 | XP_006716870.1 | 17 | 3336 | |||
XM_011517959.2 | XP_011516261.1 | 26 | 4811 | |||
XM_017014917.1 | XP_016870406.1 | 25 | 4604 | |||
XM_017014918.1 | XP_016870407.1 | 27 | 5011 | |||
XM_017014919.1 | XP_016870408.1 | 27 | 4978 | |||
XM_017014920.1 | XP_016870409.1 | 26 | 4835 | |||
GenBank mRNA | AK000627.1 | - | 4 | 799 | ||
AK000693.1 | - | 7 | 899 | |||
AK001046.1 | - | 2 | 277 | |||
AK025705.1 | - | 5 | 1717 | |||
AK125842.1 | - | 19 | 3558 | |||
AK127734.1 | - | 14 | 2972 | |||
AK299842.1 | - | 12 | 2617 | |||
AK302901.1 | - | 7 | 2028 | |||
AL834521.1 | - | 4 | 971 | |||
BC144484.1 | - | 22 | 4418 | |||
BC144485.1 | - | 21 | 3794 | |||
CR627367.1 | - | 6 | 1902 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
