siRNA Details
Gene Symbol: DENND4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017925.6 | NP_060395.5 | 26 | 4831 | ||
NR_073201.3 | 26 | 4697 | ||||
XM_006716804.3 | XP_006716867.1 | 27 | 4946 | |||
XM_006716805.3 | XP_006716868.1 | 27 | 4946 | |||
XM_006716807.3 | XP_006716870.1 | 18 | 3471 | |||
XM_017014917.1 | XP_016870406.1 | 26 | 4739 | |||
XM_017014918.1 | XP_016870407.1 | 28 | 5146 | |||
XM_017014919.1 | XP_016870408.1 | 28 | 5113 | |||
XM_017014920.1 | XP_016870409.1 | 27 | 4970 | |||
GenBank mRNA | AK000627.1 | - | 5 | 934 | ||
AK000693.1 | - | 8 | 1034 | |||
AK001046.1 | - | 3 | 412 | |||
AK025705.1 | - | 6 | 1852 | |||
AK125842.1 | - | 20 | 3693 | |||
AK127734.1 | - | 15 | 3107 | |||
AK299842.1 | - | 13 | 2752 | |||
AK302901.1 | - | 8 | 2163 | |||
AL834521.1 | - | 5 | 1106 | |||
BC144484.1 | - | 23 | 4553 | |||
BC144485.1 | - | 22 | 3929 | |||
CR627367.1 | - | 7 | 2037 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
