siRNA Details
Gene Symbol: DENND4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017925.6 | NP_060395.5 | 29 | 5401 | ||
NR_073201.3 | 29 | 5267 | ||||
XM_006716804.3 | XP_006716867.1 | 30 | 5516 | |||
XM_006716805.3 | XP_006716868.1 | 30 | 5516 | |||
XM_006716807.3 | XP_006716870.1 | 21 | 4041 | |||
XM_017014917.1 | XP_016870406.1 | 29 | 5309 | |||
XM_017014918.1 | XP_016870407.1 | 31 | 5716 | |||
XM_017014919.1 | XP_016870408.1 | 31 | 5683 | |||
XM_017014920.1 | XP_016870409.1 | 30 | 5540 | |||
GenBank mRNA | AK000693.1 | - | 11 | 1604 | ||
AK001046.1 | - | 6 | 982 | |||
AK025705.1 | - | 9 | 2422 | |||
AK125842.1 | - | 23 | 4263 | |||
AK127734.1 | - | 18 | 3677 | |||
AK299842.1 | - | 16 | 3322 | |||
AK302901.1 | - | 11 | 2733 | |||
AL834521.1 | - | 8 | 1676 | |||
BC144484.1 | - | 26 | 5123 | |||
BC144485.1 | - | 25 | 4499 | |||
CR627367.1 | - | 10 | 2607 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
