siRNA Details
Gene Symbol: ZNF429 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001415.2 | NP_001001415.2 | 4 | 1032 | ||
XM_017026748.1 | XP_016882237.1 | 5 | 1531 | |||
XM_017026749.1 | XP_016882238.1 | 5 | 3445 | |||
XM_017026750.1 | XP_016882239.1 | 5 | 1635 | |||
XM_017026751.1 | XP_016882240.1 | 4 | 1546 | |||
XM_017026752.1 | XP_016882241.1 | 4 | 1088 | |||
XM_017026753.1 | XP_016882242.1 | 6 | 1384 | |||
XM_017026754.1 | XP_016882243.1 | 5 | 1329 | |||
XM_017026755.1 | XP_016882244.1 | 6 | 3468 | |||
XM_017026756.1 | XP_016882245.1 | 5 | 1336 | |||
XM_017026757.1 | XP_016882246.1 | 3 | 1282 | |||
GenBank mRNA | AF325191.1 | - | 5 | 924 | ||
AY269786.1 | - | 5 | 924 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
