siRNA Details
Gene Symbol: ZNF429 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001415.2 | NP_001001415.2 | 4 | 1628 | ||
XM_017026748.1 | XP_016882237.1 | 5 | 2127 | |||
XM_017026749.1 | XP_016882238.1 | 5 | 4041 | |||
XM_017026750.1 | XP_016882239.1 | 5 | 2231 | |||
XM_017026751.1 | XP_016882240.1 | 4 | 2142 | |||
XM_017026752.1 | XP_016882241.1 | 4 | 1684 | |||
XM_017026753.1 | XP_016882242.1 | 6 | 1980 | |||
XM_017026754.1 | XP_016882243.1 | 5 | 1925 | |||
XM_017026755.1 | XP_016882244.1 | 6 | 4064 | |||
XM_017026756.1 | XP_016882245.1 | 5 | 1932 | |||
XM_017026757.1 | XP_016882246.1 | 3 | 1878 | |||
GenBank mRNA | AY269786.1 | - | 5 | 1520 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
