siRNA Details
Gene Symbol: NELFE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002904.5 | NP_002895.3 | 7 | 654 | ||
XM_006715205.3 | XP_006715268.1 | 7 | 523 | |||
XM_011514913.2 | XP_011513215.1 | 7 | 594 | |||
XM_017011299.1 | XP_016866788.1 | 7 | 579 | |||
GenBank mRNA | AK300898.1 | - | 7 | 529 | ||
AK302652.1 | - | 7 | 641 | |||
AK303152.1 | - | 6 | 701 | |||
BC025235.1 | - | 7 | 505 | 4876045 | ||
BC050617.2 | - | 7 | 521 | 6168937 | ||
JQ313904.1 | - | 6 | 450 | |||
L03411.1 | - | 7 | 536 | |||
X16105.1 | - | 4 | 320 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
