siRNA Details
Gene Symbol: GOT1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152413.2 | NP_689626.2 | 6 | 731 | ||
XM_005273399.3 | XP_005273456.1 | 6 | 1100 | |||
XM_006716285.3 | XP_006716348.1 | 5 | 1011 | |||
XM_011544391.2 | XP_011542693.1 | 3 | 187 | |||
GenBank mRNA | BC029504.1 | - | 6 | 732 | 5273324 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
