siRNA Details
Gene Symbol: GOT1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152413.2 | NP_689626.2 | 6 | 759 | ||
XM_005273399.3 | XP_005273456.1 | 6 | 1128 | |||
XM_006716285.3 | XP_006716348.1 | 5 | 1039 | |||
XM_011544391.2 | XP_011542693.1 | 3 | 215 | |||
GenBank mRNA | BM559469.1 | - | 760 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
