siRNA Details
Gene Symbol: SEC16B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033127.2 | NP_149118.2 | 16 | 2056 | ||
GenBank mRNA | AB063357.1 | - | 16 | 2056 | ||
AK090971.1 | - | 2 | 158 | |||
AK098627.1 | - | 5 | 676 | |||
AK314585.1 | - | 15 | 2060 | |||
EF125213.1 | - | 15 | 1966 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
