siRNA Details
Gene Symbol: SEC16B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033127.2 | NP_149118.2 | 17 | 2160 | ||
GenBank mRNA | AB063357.1 | - | 17 | 2160 | ||
AK090971.1 | - | 3 | 262 | |||
AK098627.1 | - | 6 | 780 | |||
AK314585.1 | - | 16 | 2164 | |||
EF125213.1 | - | 16 | 2070 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
