siRNA Details
Gene Symbol: MS4A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206893.3 | NP_996776.2 | 8 | 1460 | ||
GenBank mRNA | AK122633.1 | - | 8 | 1456 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
