siRNA Details
Gene Symbol: WDR49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178824.3 | NP_849146.1 | 2 | 398 | ||
GenBank mRNA | AK097556.1 | - | 6 | 1296 | ||
AK294387.1 | - | 2 | 286 | |||
BC035512.2 | - | 2 | 398 | 5199615 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
