siRNA Details
Gene Symbol: SPATA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139073.3 | NP_620712.2 | 1 | 301 | ||
XM_005246303.2 | XP_005246360.1 | 1 | 299 | |||
XM_017003363.1 | XP_016858852.1 | 1 | 299 | |||
GenBank mRNA | AK093385.1 | - | 1 | 299 | ||
AK126086.1 | - | 2 | 247 | |||
AY032925.1 | - | 2 | 399 | |||
BC047704.1 | - | 2 | 244 | 5744971 | ||
DB034232.1 | - | 1 | 299 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
