siRNA Details
Gene Symbol: TBC1D31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145088.1 | NP_001138560.1 | 6 | 876 | ||
NM_145647.3 | NP_663622.2 | 6 | 876 | |||
XM_005251102.1 | XP_005251159.1 | 6 | 876 | |||
XM_005251103.1 | XP_005251160.1 | 6 | 876 | |||
XM_005251104.1 | XP_005251161.1 | 6 | 876 | |||
XM_011517378.2 | XP_011515680.1 | 6 | 1391 | |||
XM_011517379.2 | XP_011515681.1 | 6 | 876 | |||
XM_017013983.1 | XP_016869472.1 | 6 | 876 | |||
XM_017013984.1 | XP_016869473.1 | 5 | 998 | |||
XM_017013985.1 | XP_016869474.1 | 5 | 871 | |||
XM_017013986.1 | XP_016869475.1 | 5 | 732 | |||
XM_017013987.1 | XP_016869476.1 | 6 | 876 | |||
XM_017013988.1 | XP_016869477.1 | 5 | 711 | |||
XM_017013989.1 | XP_016869478.1 | 5 | 874 | |||
XM_017013990.1 | XP_016869479.1 | 5 | 674 | |||
GenBank mRNA | AK056434.1 | - | 6 | 877 | ||
AK092041.2 | - | 3 | 2100 | |||
AK094612.1 | - | 5 | 674 | |||
BC026969.1 | - | 2 | 197 | |||
BC101724.1 | - | 6 | 843 | 8069230 | ||
BC105800.1 | - | 6 | 837 | |||
BC111954.1 | - | 6 | 843 | 8327422 | ||
BC143523.1 | - | 5 | 727 | |||
BC143525.1 | - | 6 | 843 | 9052034 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
