siRNA Details
Gene Symbol: PIEZO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022068.3 | NP_071351.2 | 21 | 3262 | ||
XM_011525723.2 | XP_011524025.1 | 24 | 4192 | |||
XM_011525724.2 | XP_011524026.1 | 23 | 4135 | |||
XM_011525725.2 | XP_011524027.1 | 22 | 4102 | |||
XM_011525726.2 | XP_011524028.1 | 24 | 4192 | |||
XM_017025918.1 | XP_016881407.1 | 23 | 4153 | |||
GenBank mRNA | AK127627.1 | - | 4 | 552 | ||
JN790819.1 | - | 21 | 3096 |

More Information
Other Species Targeted

Gene Ontology Categories:
