siRNA Details
Gene Symbol: PARP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017554.2 | NP_060024.2 | 14 | 4338 | ||
XM_011512928.2 | XP_011511230.1 | 14 | 4429 | |||
GenBank mRNA | AB033094.1 | - | 9 | 2308 | ||
AK001770.1 | - | 10 | 1366 | |||
AK022542.1 | - | 2 | 143 | |||
AK026003.1 | - | 9 | 2167 | |||
AK304269.1 | - | 14 | 4306 | |||
AY134858.1 | - | 9 | 3511 | |||
BC032804.1 | - | 1 | 54 | |||
BX648758.1 | - | 10 | 3656 | |||
DQ063584.1 | - | 11 | 3802 | |||
DQ063585.1 | - | 11 | 3802 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
