siRNA Details
Gene Symbol: PARP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017554.2 | NP_060024.2 | 15 | 4902 | ||
XM_011512928.2 | XP_011511230.1 | 15 | 4993 | |||
GenBank mRNA | AB033094.1 | - | 10 | 2872 | ||
AK022542.1 | - | 3 | 707 | |||
AK026003.1 | - | 112 | ||||
AY134858.1 | - | 10 | 4075 | |||
BC032804.1 | - | 2 | 618 | |||
BX648758.1 | - | 11 | 4220 | |||
DQ063584.1 | - | 12 | 4366 | |||
DQ063585.1 | - | 12 | 4366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
