siRNA Details
Gene Symbol: KIF25-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103750.1 | 3 | 894 | |||
GenBank mRNA | AB016899.1 | - | 2 | 1230 | ||
AB016900.1 | - | 3 | 894 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
