siRNA Details
Gene Symbol: TP53AIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195194.1 | NP_001182123.1 | 1 | 31 | ||
NM_001195195.1 | NP_001182124.1 | 2 | 697 | |||
NM_001251964.1 | NP_001238893.1 | 2 | 697 | |||
NM_022112.2 | NP_071395.2 | 2 | 697 | |||
XM_017018113.1 | XP_016873602.1 | 2 | 1462 | |||
XM_017018114.1 | XP_016873603.1 | 2 | 1468 | |||
XM_017018115.1 | XP_016873604.1 | 2 | 1471 | |||
XM_017018116.1 | XP_016873605.1 | 2 | 4727 | |||
XM_017018117.1 | XP_016873606.1 | 2 | 1467 | |||
GenBank mRNA | AB045830.1 | - | 2 | 225 | ||
AB045831.1 | - | 2 | 225 | |||
AB045832.1 | - | 2 | 225 | |||
AK310612.1 | - | 2 | 383 | |||
AK314286.1 | - | 2 | 441 | |||
AY302135.1 | - | 1 | 15 | |||
DB111427.1 | - | 2 | 441 | |||
DB129822.1 | - | 2 | 383 | |||
EF693749.1 | - | 2 | 141 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
