siRNA Details
Gene Symbol: ANKRD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286721.1 | NP_001273650.1 | 5 | 912 | ||
NM_017664.3 | NP_060134.2 | 4 | 818 | |||
NR_104586.1 | 5 | 950 | ||||
NR_104587.1 | 5 | 912 | ||||
GenBank mRNA | AK094222.1 | - | 1 | 818 | ||
AK095763.1 | - | 2 | 345 | |||
BC001727.1 | - | 5 | 811 | 3354202 | ||
BC039715.1 | - | 5 | 880 | |||
BX354283.2 | - | 5 | 809 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
