siRNA Details
Gene Symbol: ANKRD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286721.1 | NP_001273650.1 | 5 | 886 | ||
NM_017664.3 | NP_060134.2 | 4 | 792 | |||
NR_104586.1 | 5 | 924 | ||||
NR_104587.1 | 5 | 886 | ||||
GenBank mRNA | AK000100.1 | - | 4 | 754 | ||
AK094222.1 | - | 1 | 792 | |||
AK095763.1 | - | 2 | 319 | |||
BC001727.1 | - | 5 | 785 | 3354202 | ||
BC039715.1 | - | 5 | 854 | |||
BX354283.2 | - | 5 | 783 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
