siRNA Details
Gene Symbol: COQ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015697.7 | NP_056512.5 | 5 | 894 | ||
XM_011531855.2 | XP_011530157.1 | 5 | 1105 | |||
XM_011531857.2 | XP_011530159.1 | 5 | 1103 | |||
XM_011531859.2 | XP_011530161.1 | 5 | 1102 | |||
XM_011531860.2 | XP_011530162.1 | 5 | 1102 | |||
XM_011531862.2 | XP_011530164.1 | 5 | 1101 | |||
XM_011531863.2 | XP_011530165.1 | 5 | 1098 | |||
XM_011531866.2 | XP_011530168.1 | 5 | 1094 | |||
XM_011531867.2 | XP_011530169.1 | 5 | 1017 | |||
XM_017008031.1 | XP_016863520.1 | 5 | 1017 | |||
XM_017008032.1 | XP_016863521.1 | 3 | 287 | |||
GenBank mRNA | AF091086.1 | - | 4 | 443 | ||
BQ363759.1 | - | 2 | 170 | |||
CR456860.1 | - | 2 | 153 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
