siRNA Details
Gene Symbol: COQ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015697.7 | NP_056512.5 | 4 | 765 | ||
XM_011531855.2 | XP_011530157.1 | 4 | 976 | |||
XM_011531857.2 | XP_011530159.1 | 4 | 974 | |||
XM_011531859.2 | XP_011530161.1 | 4 | 973 | |||
XM_011531860.2 | XP_011530162.1 | 4 | 973 | |||
XM_011531862.2 | XP_011530164.1 | 4 | 972 | |||
XM_011531863.2 | XP_011530165.1 | 4 | 969 | |||
XM_011531866.2 | XP_011530168.1 | 4 | 965 | |||
XM_011531867.2 | XP_011530169.1 | 4 | 888 | |||
XM_017008031.1 | XP_016863520.1 | 4 | 888 | |||
XM_017008032.1 | XP_016863521.1 | 2 | 158 | |||
GenBank mRNA | AF091086.1 | - | 3 | 314 | ||
AJ621061.3 | - | 4 | 765 | |||
AK129583.1 | - | 4 | 521 | |||
AK300972.1 | - | 4 | 646 | |||
BC008804.2 | - | 4 | 622 | 3954787 | ||
BC020728.1 | - | 4 | 662 | 4696019 | ||
BG699931.1 | - | 4 | 652 | |||
CR456860.1 | - | 1 | 24 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
