siRNA Details
Gene Symbol: WDR55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017706.4 | NP_060176.2 | 7 | 1107 | ||
XM_005268469.2 | XP_005268526.1 | 7 | 1107 | |||
XM_017009600.1 | XP_016865089.1 | 8 | 894 | |||
GenBank mRNA | AK000202.1 | - | 3 | 473 | ||
AK025355.1 | - | 7 | 1054 | |||
AK056861.1 | - | 5 | 1206 | |||
AK122862.1 | - | 5 | 2542 | |||
AK293417.1 | - | 5 | 551 | |||
BC002482.2 | - | 6 | 813 | |||
BC041687.1 | - | 7 | 989 | |||
BC068485.1 | - | 7 | 1053 | 30334628 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
