siRNA Details
Gene Symbol: WDR55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017706.4 | NP_060176.2 | 1 | 140 | ||
XM_005268469.2 | XP_005268526.1 | 1 | 140 | |||
GenBank mRNA | AK025355.1 | - | 1 | 87 | ||
AK056861.1 | - | 1 | 71 | |||
BC068485.1 | - | 1 | 86 | 30334628 | ||
BQ069026.1 | - | 1 | 111 | |||
DB210296.1 | - | 1 | 140 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
