siRNA Details
Gene Symbol: PRPF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006445.3 | NP_006436.3 | 3 | 336 | ||
GenBank mRNA | AB007510.1 | - | 3 | 263 | ||
AF092565.1 | - | 3 | 260 | |||
BC000579.2 | - | 3 | 301 | |||
BC064370.1 | - | 3 | 301 | 5587081 | ||
CD698473.1 | - | 3 | 358 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
